Merck
Gene silencing for epidermal growth factor receptor variant III induces cell-specific cytotoxicity.
Molecular cancer therapeutics (2008-11-13)
Farnaz Yamoutpour, Vidya Bodempudi, Shay E Park, Weihong Pan, Mary Jean Mauzy, Robert A Kratzke, Arkadiusz Dudek, David A Potter, Richard A Woo, Donald M O'Rourke, Donald J Tindall, Faris Farassati
RESUMEN
Epidermal growth factor receptor variant III (EGFRvIII) is a constitutively active mutant form of EGFR that is expressed in 40% to 50% of gliomas and several other malignancies. Here, we describe the therapeutic effects of silencing EGFRvIII on glioma cell lines in vitro and in vivo. A small interfering RNA molecule against EGFRvIII was introduced into EGFRvIII-expressing glioma cells (U87Delta) by electroporation resulting in complete inhibition of expression of EGFRvIII as early as 48 h post-treatment. During EGFRvIII silencing, a decrease in the proliferation and invasiveness of U87Delta cells was accompanied by an increase in apoptosis (P < 0.05). Notably, EGFRvIII silencing inhibited the signal transduction machinery downstream of EGFRvIII as evidenced by decreases in the activated levels of Ras and extracellular signal-regulated kinase. A lentivirus capable of expressing anti-EGFRvIII short hairpin RNA was also able to achieve progressive silencing of EGFRvIII in U87Delta cells in addition to inhibiting cell proliferation, invasiveness, and colony formation in a significant manner (P < 0.05). Silencing EGFRvIII in U87Delta cultures with this virus reduced the expression of factors involved in epithelial-mesenchymal transition including N-cadherin, beta-catenin, Snail, Slug, and paxillin but not E-cadherin. The anti-EGFRvIII lentivirus also affected the cell cycle progression of U87Delta cells with a decrease in G(1) and increase in S and G(2) fractions. In an in vivo model, tumor growth was completely inhibited in severe combined immunodeficient mice (n = 10) injected s.c. with U87Delta cells treated with the anti-EGFRvIII lentivirus (P = 0.005). We conclude that gene specific silencing of EGFRvIII is a promising strategy for treating cancers that contain this mutated receptor.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Kit de análisis de activación Ras
Sigma-Aldrich
Ral Activation Assay Kit, Non-radioactive Ral Activation Assay Kit can be used to precipitate Ral-A-GTP from cell lysates & detection by a Ral-A specific antibody.