Merck
  • High-Performance As-Cast Nonfullerene Polymer Solar Cells with Thicker Active Layer and Large Area Exceeding 11% Power Conversion Efficiency.
High-Performance As-Cast Nonfullerene Polymer Solar Cells with Thicker Active Layer and Large Area Exceeding 11% Power Conversion Efficiency.
Advanced materials (Deerfield Beach, Fla.) (2017-12-14)
Qunping Fan, Yan Wang, Maojie Zhang, Bo Wu, Xia Guo, Yufeng Jiang, Wanbin Li, Bing Guo, Chennan Ye, Wenyan Su, Jin Fang, Xuemei Ou, Feng Liu, Zhixiang Wei, Tze Chien Sum, Thomas P Russell, Yongfang Li
RESUMEN
In this work, a nonfullerene polymer solar cell (PSC) based on a wide bandgap polymer donor PM6 containing fluorinated thienyl benzodithiophene (BDT-2F) unit and a narrow bandgap small molecule acceptor 2,2'-((2Z,2'Z)-((4,4,9,9-tetrahexyl-4,9-dihydro-s-indaceno[1,2-b:5,6-b']dithiophene-2,7-diyl)bis(methanylylidene))bis(3-oxo-2,3-dihydro-1H-indene-2,1-diylidene))dimalononitrile (IDIC) is developed. In addition to matched energy levels and complementary absorption spectrum with IDIC, PM6 possesses high crystallinity and strong π-π stacking alignment, which are favorable to charge carrier transport and hence suppress recombination in devices. As a result, the PM6:IDIC-based PSCs without extra treatments show an outstanding power conversion efficiency (PCE) of 11.9%, which is the record value for the as-cast PSC devices reported in the literature to date. Moreover, the device performances are insensitive to the active layer thickness (≈95-255 nm) and device area (0.20-0.81 cm2 ) with PCEs of over 11%. Besides, the PM6:IDIC-based flexible PSCs with a large device area of 1.25 cm2 exhibit a high PCE of 6.54%. These results indicate that the PM6:IDIC blend is a promising candidate for future roll-to-roll mass manufacturing and practical application of highly efficient PSCs.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
PBDB-T-2F