Merck
  • Pro-opiomelanocortin and its Processing Enzymes Associate with Plaque Stability in Human Atherosclerosis - Tampere Vascular Study.
Pro-opiomelanocortin and its Processing Enzymes Associate with Plaque Stability in Human Atherosclerosis - Tampere Vascular Study.
Scientific reports (2018-10-12)
Petteri Rinne, Leo-Pekka Lyytikäinen, Emma Raitoharju, James J Kadiri, Ivana Kholova, Mika Kähönen, Terho Lehtimäki, Niku Oksala
RESUMEN
α-melanocyte-stimulating hormone (α-MSH) is processed from pro-opiomelanocortin (POMC) and mediates anti-inflammatory actions in leukocytes. α-MSH also promotes macrophage reverse cholesterol transport by inducing ATP-binding cassette transporters ABCA1 and ABCG1. Here we investigated the regulation of POMC and α-MSH expression in atherosclerosis. First, transcript levels of POMC and its processing enzymes were analyzed in human arterial plaques (n = 68) and non-atherosclerotic controls (n = 24) as well as in whole blood samples from coronary artery disease patients (n = 55) and controls (n = 45) by microarray. POMC expression was increased in femoral plaques compared to control samples as well as in unstable advanced plaques. α-MSH-producing enzyme, carboxypeptidase E, was down-regulated, whereas prolylcarboxypeptidase, an enzyme inactivating α-MSH, was up-regulated in unstable plaques compared to stable plaques, suggesting a possible reduction in intraplaque α-MSH levels. Second, immunohistochemical analyses revealed the presence of α-MSH in atherosclerotic plaques and its localization in macrophages and other cell types. Lastly, supporting the role of α-MSH in reverse cholesterol transport, POMC expression correlated with ABCA1 and ABCG1 in human plaque and whole blood samples. In conclusion, α-MSH is expressed in atherosclerotic plaques and its processing enzymes associate with plaque stability, suggesting that measures to enhance the local bioavailability of α-MSH might protect against atherosclerosis.