Merck
Effective critical micellar concentration of a zwitterionic detergent: a fluorimetric study on n-dodecyl phosphocholine.
Journal of fluorescence (2009-09-17)
Pasquale Palladino, Filomena Rossi, Raffaele Ragone
RESUMEN
We have investigated the effect of ionic strength on the aggregation behavior of n-dodecyl phosphocholine. On the basis of the classical Corrin-Harkins relation, the critical micellar concentration of this detergent decreases with a biphasic trend on lithium chloride addition. It is nearly constant below 150 mM salt, with a mean value of 0.91 mM, whereas it undergoes a dramatic 80-fold decrease in 7 M LiCl. Such a drop in the critical micellar concentration could be explained by the effect of salting out and the implication of phosphocholine head groups on the organization of surrounding water. Knowledge of the effective critical micellar concentration of n-dodecyl phosphocholine could be useful in the purification of membrane proteins in non-denaturing conditions.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Avanti
MAPCHO-12, Avanti Research - A Croda Brand
Avanti
MAPCHO-10, Avanti Research - A Croda Brand 850334P, powder