Merck
Nesfatin-1 ameliorates testicular injury and supports gonadal function in rats induced with testis torsion.
Peptides (2018-07-22)
Sevil Arabaci Tamer, Alper Yildirim, M Kutay Köroğlu, Özge Çevik, Feriha Ercan, Berrak Ç Yeğen
RESUMEN
Testicular torsion causes ischemia-reperfusion injury and an increased risk of infertility. Nesfatin-1 is a novel peptide with antioxidant, anti-inflammatory and anti-apoptotic properties. In the present study, we aimed to investigate the putative beneficial effects of nesfatin-1 on oxidative injury and impaired testicular function induced by testis torsion. Under anesthesia, male Sprague-Dawley rats (180-230 g; n = 24) had sham-operation or they underwent testicular torsion by rotating the left testis 720° and fixing it for 2 h, followed by a 2-h detorsion. Rats in each group were treated intraperitoneally with either nesfatin-1 (0.3 μg/kg) or saline prior to the torsion or sham-torsion. At the end of the 4-h experimental period, tissue samples were removed for evaluation of spermatozoa, molecular and histochemical analyses. In saline-treated torsion/detorsion group, a high percentage of abnormal spermatozoa with head defects was observed, which was abolished in nesfatin-1-treated torsion/detorsion group. The levels of 8-OHdG, tumor necrosis factor (TNF)-alpha, interleukin (IL)-6, caspase-3 were increased in the saline-treated torsion/detorsion group as compared to sham-operated group, while nesfatin-1 pre-treatment significantly decreased the expressions of the pro-inflammatory cytokines, depressed apoptosis, and also reduced the tubular degeneration. In addition, nesfatin-1 in torsion/detorsion group elevated expressions of transforming growth factor (TGF)-beta and reduced expressions of protein kinase B (AKT) and cAMP response element binding protein (CREB) in the testis tissue. The present findings show that nesfatin-1, by regulating AKT and CREB signaling pathways and pro-inflammatory/anti-inflammatory cytokine balance, preserves the spermatogenic cells and ameliorates torsion-detorsion-induced tubular degeneration.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Millipore
Skim Milk Powder, suitable for microbiology