Merck
Non-inverted pedicle internal limiting membrane transposition for large macular holes.
Eye (London, England) (2018-05-31)
Zizhong Hu, Xiaojian Ye, Xuehua Lv, Kang Liang, Weiwei Zhang, Xi Chen, Erbing Cao, Xunyi Gu, Qinghuai Liu, Ping Xie
RESUMEN
The purpose of this study is to investigate the effectiveness of a new surgical technique of non-inverted pedicle internal limiting membrane (ILM) transposition for the treatment of eyes with large macular hole. This is a retrospective, consecutive, interventional case series. Twelve eyes of 12 consecutive patients who underwent vitrectomy for the treatment of a large macular hole (MH size > 400 µm) were treated. ILM was peeled and left with a pedicle attached to the superior temporal retina. The macular hole was covered by transposition of the pedicle ILM in a non-inverted way. Preoperative and postoperative best-corrected visual acuity (BCVA), SD-OCT image, macular sensitivity by microperimetry, and multifocal electroretinogram (mERG) response were evaluated. All of the patients were followed for more than 3 months. Postoperative OCT examination confirmed 11 of 12 macular hole closed (91.7%). Six macular hole filled with silicone oil closed as early as the next day. The postoperative BCVA significantly increased compared with preoperative BCVA (P = 0.002). The improvement of macular sensitivity within 2° and 8° circle was also statistical significant (P = 0.018 and P = 0.017, respectively). Fixation stability, shown as the percentage of fixation point within the 2° circle and 4° circle, was remarkably improved (P = 0.017 and P = 0.018, respectively). The R1/R2 and R1/R4 ring ratios also increased significantly as compared with that of baseline. These findings indicate that the non-inverted pedicle ILM transposition results in a high incidence of anatomic closure with good visual outcome for the treatment of large macular hole.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Tetrahydropyran, anhydrous, 99%
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad