Merck
X-linked IAP is a direct inhibitor of cell-death proteases.
Nature (1997-07-17)
Q L Deveraux, R Takahashi, G S Salvesen, J C Reed
RESUMEN
The inhibitor-of-apoptosis (IAP) family of genes has an evolutionarily conserved role in regulating programmed cell death in animals ranging from insects to humans. Ectopic expression of human IAP proteins can suppress cell death induced by a variety of stimuli, but the mechanism of this inhibition was previously unknown. Here we show that human X-chromosome-linked IAP directly inhibits at least two members of the caspase family of cell-death proteases, caspase-3 and caspase-7. As the caspases are highly conserved throughout the animal kingdom and are the principal effectors of apoptosis, our findings suggest how IAPs might inhibit cell death, providing evidence for a mechanism of action for these mammalian cell-death suppressors.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
XIAP, GST tagged human, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution