Merck
  • Antimicrobial Activity of Dalbavancin against Staphylococcus aureus with Decreased Susceptibility to Glycopeptides, Daptomycin, and/or Linezolid from U.S. Medical Centers.
Antimicrobial Activity of Dalbavancin against Staphylococcus aureus with Decreased Susceptibility to Glycopeptides, Daptomycin, and/or Linezolid from U.S. Medical Centers.
Antimicrobial agents and chemotherapy (2017-12-22)
Helio S Sader, Rodrigo E Mendes, Leonard R Duncan, Michael A Pfaller, Robert K Flamm
RESUMEN
Dalbavancin activity was assessed against a large collection of Staphylococcus aureus isolates (n = 59,903), including isolates with decreased susceptibility to vancomycin (MIC, ≥2 mg/liter; n = 1,141), daptomycin (MIC, ≥2 mg/liter; n = 48), telavancin (MIC, ≥0.12 mg/liter; n = 52), teicoplanin (MIC, ≥4 mg/liter; n = 143), and/or linezolid (MIC, ≥8 mg/liter; n = 25). Dalbavancin displayed susceptibility rates ranging from 90.4% (isolates with telavancin MIC ≥0.12 mg/liter) to 100.0% (linezolid-resistant isolates) and lower MIC values than the comparators against these resistant subsets.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Dalbavancin, ≥90% (HPLC)