Merck
Genetic instability triggered by G-quadruplex interacting Phen-DC compounds in Saccharomyces cerevisiae.
Nucleic acids research (2010-03-13)
Aurèle Piazza, Jean-Baptiste Boulé, Judith Lopes, Katie Mingo, Eric Largy, Marie-Paule Teulade-Fichou, Alain Nicolas
RESUMEN
G-quadruplexes are nucleic acid secondary structures for which many biological roles have been proposed but whose existence in vivo has remained elusive. To assess their formation, highly specific G-quadruplex ligands are needed. Here, we tested Phen-DC(3) and Phen-DC(6), two recently released ligands of the bisquinolinium class. In vitro, both compounds exhibit high affinity for the G4 formed by the human minisatellite CEB1 and inhibit efficiently their unwinding by the yeast Pif1 helicase. In vivo, both compounds rapidly induced recombination-dependent rearrangements of CEB1 inserted in the Saccharomyces cerevisiae genome, but did not affect the stability of other tandem repeats lacking G-quadruplex forming sequences. The rearrangements yielded simple-deletion, double-deletion or complex reshuffling of the polymorphic motif units, mimicking the phenotype of the Pif1 inactivation. Treatment of Pif1-deficient cells with the Phen-DC compounds further increased CEB1 instability, revealing additional G4 formation per cell. In sharp contrast, the commonly used N-methyl-mesoporphyrin IX G-quadruplex ligand did not affect CEB1 stability. Altogether, these results demonstrate that the Phen-DC bisquinolinium compounds are potent molecular tools for probing the formation of G-quadruplexes in vivo, interfere with their processing and elucidate their biological roles.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
PhenDC3, ≥97% (HPLC)