Merck
Structure-activity relationship study of EphB3 receptor tyrosine kinase inhibitors.
Bioorganic & medicinal chemistry letters (2009-09-29)
Lixin Qiao, Sungwoon Choi, April Case, Thomas G Gainer, Kathleen Seyb, Marcie A Glicksman, Donald C Lo, Ross L Stein, Gregory D Cuny
RESUMEN
A structure-activity relationship study for a 2-chloroanilide derivative of pyrazolo[1,5-a]pyridine revealed that increased EphB3 kinase inhibitory activity could be accomplished by retaining the 2-chloroanilide and introducing a phenyl or small electron donating substituents to the 5-position of the pyrazolo[1,5-a]pyridine. In addition, replacement of the pyrazolo[1,5-a]pyridine with imidazo[1,2-a]pyridine was well tolerated and resulted in enhanced mouse liver microsome stability. The structure-activity relationship for EphB3 inhibition of both heterocyclic series was similar. Kinase inhibitory activity was also demonstrated for representative analogs in cell culture. An analog (32, LDN-211904) was also profiled for inhibitory activity against a panel of 288 kinases and found to be quite selective for tyrosine kinases. Overall, these studies provide useful molecular probes for examining the in vitro, cellular and potentially in vivo kinase-dependent function of EphB3 receptor.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
5-Bromo-2-nitropyridine, 99%