Merck
  • Rapid synthesis of maleimide functionalized fluorine-18 labeled prosthetic group using "radio-fluorination on the Sep-Pak" method.
Rapid synthesis of maleimide functionalized fluorine-18 labeled prosthetic group using "radio-fluorination on the Sep-Pak" method.
Journal of labelled compounds & radiopharmaceuticals (2018-03-27)
Falguni Basuli, Xiang Zhang, Elaine M Jagoda, Peter L Choyke, Rolf E Swenson
RESUMEN
Following our recently published fluorine-18 labeling method, "Radio-fluorination on the Sep-Pak", we have successfully synthesized 6-[18 F]fluoronicotinaldehyde by passing a solution (1:4 acetonitrile: t-butanol) of its quaternary ammonium salt precursor, 6-(N,N,N-trimethylamino)nicotinaldehyde trifluoromethanesulfonate (2), through a fluorine-18 containing anion exchange cartridge (PS-HCO3 ). Over 80% radiochemical conversion was observed using 10 mg of precursor within 1 minute. The [18 F]fluoronicotinaldehyde ([18 F]5) was then conjugated with 1-(6-(aminooxy)hexyl)-1H-pyrrole-2,5-dione to prepare the fluorine-18 labeled maleimide functionalized prosthetic group, 6-[18 F]fluoronicotinaldehyde O-(6-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexyl) oxime, 6-[18 F]FPyMHO ([18 F]6). The current Sep-Pak method not only improves the overall radiochemical yield (50 ± 9%, decay-corrected, n = 9) but also significantly reduces the synthesis time (from 60-90 minutes to 30 minutes) when compared with literature methods for the synthesis of similar prosthetic groups.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
6-Fluoro-2-pyridinecarbaldehyde