Merck
Fluorenone based fluorescent probe for selective "turn-on" detection of pyrophosphate and alanine.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2017-11-21)
Daniel Thangadurai T, Nithya I, Manjubaashini N, Bhuvanesh N, Bharathi G, Nandhakumar R, Nataraj D
RESUMEN
To sense biologically important entities with different size and dimensions, a fluorenone based fluorescent receptor was designed and synthesized. Probe 1 displayed a distinct fluorescence enhancement emission at 565nm for pyrophosphate and 530nm for alanine in polar solvent. The fluorescence titration experiments confirm 1:1 stoichiometric ratio with high-binding constant and very low limit of detection (LoD) values. Receptor 1 showed a highly selective and sensitive recognition to HP2O73- and to alanine over other competitive anions and amino acids. In addition, the fluorescence lifetime measurement and reversible binding study results support the practical importance of 1.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
1-Amino-9-fluorenone, 97%