Merck
  • Drosophila larvae fed palm fruit juice (PFJ) delay pupation via expression regulation of hormetic stress response genes linked to ageing and longevity.
Drosophila larvae fed palm fruit juice (PFJ) delay pupation via expression regulation of hormetic stress response genes linked to ageing and longevity.
Experimental gerontology (2018-03-20)
Soon-Sen Leow, Alice Luu, Swechhya Shrestha, K C Hayes, Ravigadevi Sambanthamurthi
RESUMEN
Palm fruit juice (PFJ) containing oil palm phenolics is obtained as a by-product from oil palm (Elaeis guineensis) fruit milling. It contains shikimic acid, soluble fibre and various phenolic acids including p-hydroxybenzoic acid and three caffeoylshikimic acid isomers. PFJ has also demonstrated beneficial health properties in various biological models. Increasing concentrations of PFJ and different PFJ fractions were used to assess growth dynamics and possible anti-ageing properties in fruit flies (Drosophila melanogaster) genotype w1118. Microarray gene expression analysis was performed on whole fruit fly larvae and their fat bodies, after the larvae were fed a control Standard Brandeis Diet (SBD) with or without PFJ. Transcripts from Affymetrix GeneChips were utilised to identify the possible mechanisms involved, with genes having fold changes > |1.30| and p < 0.05 considered differentially expressed. PFJ dose-dependently delayed larval growth and pupation, but not percent eclosion from pupae. Eclosed male fruit flies fed PFJ or its fractions during the larval stage tended to have 20-40% improved survival ratings over controls when allowed to age on the control diet (SBD). Microarray analysis of whole fruit fly larvae revealed that 127 genes were up-regulated, while 67 were down-regulated by PFJ. Functional analysis revealed transport and metabolic processes were up-regulated, while development and morphogenesis processes, including the nutrient-sensing Tor gene, were down-regulated by PFJ, whereas microarray analysis of larval fat bodies found 161 genes were up-regulated, while 84 genes were down-regulated. Genes involved in defence response and determination of adult lifespan, including those encoding various heat shock proteins and the antioxidant enzyme Sod2, were up-regulated, while cell cycle and growth genes were down-regulated. Thus, PFJ supplementation lengthened the growth stages in fruit fly larvae that was reflected in extended ageing of adult flies, suggesting that larval expression of hormetic stress response genes was linked to subsequent ageing and longevity.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Prunetin, ≥98.0% (TLC)