Key Documents
SU860014
Supelco
Crimp Seal with PTFE/butyl septum
gold steel seal (magnetic), open center, 8mm opening, seal diam. × total thickness × PTFE thickness 20 mm × 3.0 mm × 5 mil, PTFE/butyl rubber septum
About This Item
UNSPSC Code:
41121806
material
PTFE/butyl rubber septum
gold steel seal (magnetic)
feature
open center 8mm opening
hardness
50 (Durometer: shore A)
packaging
pkg of 100 ea
seal diam. × total thickness × PTFE thickness
20 mm × 3.0 mm × 5 mil
compatibility
for use with CPI CombiPAL
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)