Documentos
509736
Jeringas SGE para autosistema PerkinElmer®
volume 5 μL, 5F-PE-0.47, needle size 26 ga, needle L 70 mm
About This Item
UNSPSC Code:
41122001
material
cone tip stainless steel needle (point style 1)
needle size
26 ga
description
color (lime)
liquid syringe, fixed needle
feature
needle type LTN (fixed)
packaging
pkg of 1 ea
manufacturer/tradename
SGE 001953
needle L
70 mm
volume
5 μL
Legal Information
PerkinElmer is a registered trademark of PerkinElmer Inc.
Choose from one of the most recent versions:
Certificados de análisis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Atención al cliente