Documentos clave
AXYPDW500CSI
Deep well plate
for SBS FootPrint, individually wrapped, size 96 wells, size 600 μL, 25 ea of, sterile
Sinónimos:
Corning cell culture plate
About This Item
UNSPSC Code:
41121800
NACRES:
NB.15
material:
conical bottom wells
size:
600 μL
96 wells
En este momento no podemos mostrarle ni los precios ni la disponibilidad
material
conical bottom wells
sterility
sterile
manufacturer/tradename
Axygen Inc P-DW-500-C-SI
Corning
packaging
25 ea of
size
600 μL
96 wells
General description
  • Axygen® deep well assay plates are ideal for use in applications including sample collection, in vitro growth chambers and long-term storage.
  • Chemically resistant and withstands temperatures down to -80°C.
  • Ultra-low profile design for reduced space requirements.
  • Flat surface for efficient sealing using heat sealing film.
Legal Information
Axygen is a registered trademark of Corning, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)