Merck
69140
Sigma-Aldrich
Methyl Red sodium salt
ACS reagent, indicator (pH 4.2-6.2)
Sinónimos:
4-Dimethylaminoazobenzene-2′-carboxylic acid sodium salt, Acid Red 2
About This Item
Fórmula empírica (notación de Hill):
C15H14N3NaO2
Número de CAS:
Peso molecular:
291.28
Colour Index Number:
13020
Beilstein/REAXYS Number:
4065488
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
grade
ACS reagent
indicator (pH 4.2-6.2)
form
powder
technique(s)
titration: suitable
visual transition interval
4.2-6.2, pink to yellow
solubility
H2O: soluble
SMILES string
[Na+].CN(C)c1ccc(cc1)\N=N\c2ccccc2C([O-])=O
InChI
1S/C15H15N3O2.Na/c1-18(2)12-9-7-11(8-10-12)16-17-14-6-4-3-5-13(14)15(19)20;/h3-10H,1-2H3,(H,19,20);/q;+1/p-1/b17-16+;
InChI key
GNTPCYMJCJNRQB-CMBBICFISA-M
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
K Danielle Kelley et al.
Biochemistry, 49(26), 5405-5407 (2010-06-15)
It is shown that Methyl Red can be used as an indicator dye that changes color in Escherichia coli culture as a result of time- and cell density-dependent bleaching by azoreductase produced by the bacteria. For cell cultures that are
Oana Alexandru et al.
Journal of neuro-oncology, 102(1), 9-18 (2010-07-17)
A major focus of brain cancer research today is to translate understanding of glioma biology into advances in treatment, by exploring the potential of target therapy. Here we investigated the ability of three compounds belonging to the chemical class of
Chan-Ju Wang et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 66(Pt 1), 2-7 (2010-01-09)
Azoreductase 1 from Pseudomonas aeruginosa strain PAO1 (paAzoR1) catalyses the activation of the prodrug balsalazide and reduces the azo dye methyl red using reduced nicotinamide adenine dinucleotide cofactor as an electron donor. To investigate the mechanism of the enzyme, a
Z Ioannou et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 67(5), 1129-1136 (2013-02-19)
Zeolite, hematite, modified zeolite and commercial activated charcoal were examined for their ability to remove methylene blue (MB) and methyl red (MR) from their aqueous solutions. Modified zeolite and hematite were produced according to the Schwertmann and Cornell method while
Jinhui Feng et al.
Archives of biochemistry and biophysics, 520(2), 99-107 (2012-03-06)
AzoA from Enterococcus faecalis is a member of the polymeric flavin-dependent NADH-preferred azoreductase group. Little is known about the binding and interaction of NADH and azo dye in the azoreductase group. A synergetic strategy based on computational prediction, reverse genetics