Merck
31489
Supelco
Bioallethrin
PESTANAL®, analytical standard
About This Item
Fórmula empírica (notación de Hill):
C19H26O3
Número de CAS:
Peso molecular:
302.41
Beilstein/REAXYS Number:
2294836
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
grade
analytical standard
Quality Level
product line
PESTANAL®
shelf life
limited shelf life, expiry date on the label
storage condition
under inert gas (argon)
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
agriculture
environmental
format
neat
storage temp.
2-8°C
SMILES string
C\C(C)=C\C1C(C(=O)OC2CC(=O)C(CC=C)=C2C)C1(C)C
InChI
1S/C19H26O3/c1-7-8-13-12(4)16(10-15(13)20)22-18(21)17-14(9-11(2)3)19(17,5)6/h7,9,14,16-17H,1,8,10H2,2-6H3
InChI key
ZCVAOQKBXKSDMS-UHFFFAOYSA-N
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Caution
Light sensitive.
Legal Information
PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany
pictograms
Exclamation markEnvironment
signalword
Warning
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Los clientes también vieron
Andrew Kroetsch et al.
mAbs, 11(2), 411-421 (2018-12-12)
A new modality in antibody engineering has emerged in which the antigen affinity is designed to be pH dependent (PHD). In particular, combining high affinity binding at neutral pH with low affinity binding at acidic pH leads to a novel
Golla Madhubabu et al.
Inhalation toxicology, 24(3), 143-152 (2012-02-24)
Continuous inhalation of allethrin-based mosquito coil smoke may affect fertility, an aspect that has not received much attention. In this study, we attempt to understand the harmful effects on the male reproductive system caused by continuous exposure to allethrin-based mosquito
Koen Desmet et al.
Chemosphere, 71(11), 2193-2198 (2008-04-02)
Spatial concentration distribution of a chemical in an indoor environment is an important factor in the evaluation of chemical nuisances. However, straightforward techniques for the determination of this distribution are not very common and usually limited in their application. Sorptive
April P Neal et al.
Toxicological sciences : an official journal of the Society of Toxicology, 116(2), 604-613 (2010-05-15)
Pyrethroid insecticides are one of the most widely used classes of insecticides. Previous studies revealed that pyrethroids potently affect the insect voltage-gated sodium (Na(+)) channel (VGSC), resulting in prolonged channel open time. However, recent findings have suggested that pyrethroids may
Mahmut Selvi et al.
Environmental toxicology, 28(11), 644-651 (2011-09-14)
Esbiothrin, synthetic pyrethroid with quick activity against insects, is widely used against household pests and in public health. Despite widespread use, data on ecotoxicity and genotoxic effects are extremely scarce. The aim of the present study is to evaluate the