Documentos
Z558923
Sigma-Aldrich
Sigma-Aldrich® anti-static disposable unisex lab coat
light blue, size S
About This Item
UNSPSC Code:
46181532
NACRES:
NB.35
size
S
availability
not available in EU
color
light blue
General description
DL4260 Disposable Lab Coats. These tough, lightweight, full-length lab coats are splash/spray resistant and offer superior comfort/breathability, and are anti-static treated.
Details:
  • Material: SMS
  • Weight: 30 grams
  • Collar: knit
  • Pockets: 3
  • Cuffs, Heavy Duty Snaps, Latex Free: yes
Legal Information
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany