THS05117
ThalesNano CatCart® catalyst cartridge system, 30 mm L
Wilkinson′s catalyst RhCl(TPP)3
Sinónimos:
Heterogeneous catalysis system, safe Heterogeneous catalysis
About This Item
Productos recomendados
packaging
pkg of 1 ea
manufacturer/tradename
ThalesNano THS 05117
diam.
4 mm
Categorías relacionadas
General description
CatCarts are suitable for use only with Thalesnano H-Cube reactors.
Legal Information
CatCart is a registered trademark of ThalesNano Nanotechnology Inc.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c