Merck
220671
Sigma-Aldrich
L-Methionine ethyl ester hydrochloride
99%
About This Item
Fórmula lineal:
CH3SCH2CH2CH(NH2)COOC2H5 · HCl
Número de CAS:
Peso molecular:
213.73
Beilstein/REAXYS Number:
3913812
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22
assay
99%
form
powder
optical activity
[α]25/D +21°, c = 2 in ethanol
reaction suitability
reaction type: solution phase peptide synthesis
mp
90-92 °C (lit.)
application(s)
peptide synthesis
SMILES string
Cl[H].CCOC(=O)[C@@H](N)CCSC
InChI
1S/C7H15NO2S.ClH/c1-3-10-7(9)6(8)4-5-11-2;/h6H,3-5,8H2,1-2H3;1H/t6-;/m0./s1
InChI key
KPWCQEUBJAIORR-RGMNGODLSA-N
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Visite la Librería de documentos
Studies on methionine derivatives as possible sources of protected methionine in ruminant rations.
J A Ayoade et al.
Journal of the science of food and agriculture, 33(10), 949-956 (1982-10-01)
Shaikh A Ali et al.
Journal of hazardous materials, 350, 169-179 (2018-02-27)
Cyclopolymerization of N,N-diallylmethionine hydrochloride, derived from the biogenic amino acid methionine, (90 mol%) and cross-linker tetraallylpiperazinium dichloride (10 mol%) in presence of an azo-initiator afforded pH-responsive cross-linked polyzwitterion (CPZ). The structural morphology of the resin (i.e. CPZ) was examined by the BET
K Eszter Borbas et al.
Bioconjugate chemistry, 18(4), 1205-1212 (2007-06-23)
Targeted radiopharmaceuticals offer the possibility of improved tumor imaging and radiotherapy, with reduced side effects. A variety of monoclonal antibodies and antibody fragments have previously been successfully radiolabeled and used in diagnostic imaging and targeted radiotherapy of cancer. Many such
Griet Van Zeebroeck et al.
Nature chemical biology, 5(1), 45-52 (2008-12-09)
Transporter-related nutrient sensors, called transceptors, mediate nutrient activation of signaling pathways through the plasma membrane. The mechanism of action of transporting and nontransporting transceptors is unknown. We have screened 319 amino acid analogs to identify compounds that act on Gap1