HYCON® Contact Slides
HYCON® Contact Slides - Ready-to-use media for microbial testing of flat and curved surfaces
For easy and flexible microbiological monitoring of surfaces. The unique design of these HYCON® contact slides allows testing on flat, curved or irregular surfaces and makes them a universal tool and ideally suited to the production environment found in food and beverage production.
- Offered as a range of total count media as well as selective media for yeasts and molds, and for coliform bacteria.
- Manufactured under controlled aseptic conditions
- Single packed strips to minimize waste
- Shelf life of up to six months to help save costs
- Room temperature storage of nonselective media giving cost savings on temperature controlled storage
- Rectangular shape (25 cm2)
Network error: Failed to fetch