Merck
HomeReaction Design & OptimizationLithium Aminoborohydride (LAB) Reagents
Lithium Aminoborohydride (LAB) Reagents
Lithium aminoborohydride (LAB) reagents are a class of selective reagents developed in the laboratory of Professor Bakthan Singaram at the University of California, Santa Cruz. These reagents have reactivity comparable to lithium aluminum hydride (LAH). However, LABs have several advantages over LAH in that they are air-stable, nonpyrophoric, thermally stable, and hydrolyze only slowly in protic solvents above pH 4. Thus, LABs can perform in air virtually all of the transformations for which LAH is commonly used, and they offer significant advantages in safety, selectivity, ease of handling, and simple work-up procedures. LABs can reduce a variety of functional groups (Scheme 1).1
Lithium aminoborohydride (LAB) reagents
Scheme 1.Lithium aminoborohydride (LAB) reagents
In the case of N-alkyl lactams with an ester group, LAB can be used at reduced temperatures to selectively reduce the ester, leaving the lactam intact. At elevated temperatures, the lactam is also reduced to form the cyclic amine (Scheme 2).2
Cyclic amine
Scheme 2.Cyclic amine
In addition to hydride transfer, LABs can transfer the amine moiety, as in the case of reaction with halopyridines (Scheme 3)3 and primary alkyl methanesulfonates (Scheme 4).4 Alkyl methanesulfonates can be reduced to the corresponding alkanes with LAB reagents if Et3B is added in substoichiometric amounts (Scheme 5).4
Halopyridines
Scheme 3.Halopyridines
Primary alkyl methanesulfonates
Scheme 4.Primary alkyl methanesulfonates
Substoichiometric amounts
Scheme 5.Substoichiometric amounts
The LAB reagents’ ability to function as both a reducing agent and amination reagent allows the researcher to perform tandem amination-reduction reactions, as in the case of reaction with 2-halobenzonitriles.5 The LAB reagent activates the halo-benzonitrile toward nucleophilic attack by the amine moiety of the LAB reagent. This protocol is particularly effective with 2-fluorobenzonitrile (Scheme 6).
2-fluorobenzonitrile
Scheme 6.2-fluorobenzonitrile
Materials
Loading
References
1.
Aminoborohydrides. 13. Facile reduction of N-alkyl lactams with 9-borabicyclo[3.3.1]nonane (9-BBN) and lithium aminoborohydrides (LAB) reagents. 2001(4):59. https://doi.org/10.3998/ark.5550190.0002.407
2.
Flaniken JM, Collins CJ, Lanz M, Singaram B. 1999. Aminoborohydrides. 11. Facile Reduction ofN-Alkyl Lactams to the Corresponding Amines Using Lithium Aminoborohydrides. Org. Lett.. 1(5):799-801. https://doi.org/10.1021/ol990145e
3.
Thomas S, Roberts S, Pasumansky L, Gamsey S, Singaram B. 2003. Aminoborohydrides 15. The First Mild and Efficient Method for Generating 2-(Dialkylamino)-pyridines from 2-Fluoropyridine. Org. Lett.. 5(21):3867-3870. https://doi.org/10.1021/ol035430j
4.
Thomas S, Huynh T, Enriquez-Rios V, Singaram B. 2001. Aminoborohydrides. 14. Lithium Aminoborohydrides in the Selective ReductionorAmination of Alkyl Methanesulfonate Esters. Org. Lett.. 3(24):3915-3918. https://doi.org/10.1021/ol0167659
5.
Thomas S, Collins CJ, Cuzens JR, Spiciarich D, Goralski CT, Singaram B. 2001. Aminoborohydrides. 12. Novel Tandem SNAr Amination?Reduction Reactions of 2-Halobenzonitriles with LithiumN,N-Dialkylaminoborohydrides. J. Org. Chem.. 66(6):1999-2004. https://doi.org/10.1021/jo001388j