Merck
HomeCross-CouplingTrost Ligands for Allylic Alkylation
Trost Ligands for Allylic Alkylation
The Tsuji Trost cycle is a synthetic methodology used in organic chemistry for the construction of complex molecules. It involves the use of a palladium-catalyzed reaction to facilitate the formation of carbon-carbon bonds through the coupling of allylic alcohols with nucleophiles. Palladium-catalyzed asymmetric allylic alkylation (AAA) is a powerful method for the efficient construction of stereogenic centers. In contrast to many other catalytic methods, AAA has the ability to form multiple types of bonds (C–C, C–O, C–S, C–N) with a single catalyst system.
Section Overview
Nucleophilic Addition
Figure 1.Nucleophilic Addition
The Trost group at Stanford University has pioneered the use of C-2 symmetric diaminocyclohexyl (DACH) ligands in AAA, allowing for the rapid synthesis of a diverse range of chiral products with a limited number of chemical transformations. Reactions are typically high-yielding, and excellent levels of enantioselectivity are observed.1
Trost Ligands
Figure 2.Trost Ligands
Carbon Nucleophiles
In early examples of this methodology, Trost and co-workers demonstrated diesters are competent nucleophiles for the deracemization of cyclic allylic acetates, to afford chiral malonate derivatives. Since that time, soft carbon nucleophiles such as barbituric acid derivatives, β-keto esters, nitro compounds, and many others have been employed in AAA for assembly of tertiary and quaternary asymmetric centers.
Malonate Nucleophiles
Figure 3.Malonate Nucleophiles
keto
Figure 4.β-Keto Ester Nucleophiles
Oxygen Nucleophiles
Carbon-oxygen bond-forming reactions using palladium-catalyzed asymmetric allylic alkylation have been well-demonstrated in numerous natural product syntheses. Alcohols, carboxylates, and hydrogencarbonates have all been employed as O-nucleophiles.
Alcohol Nucleophiles
Figure 5.Alcohol Nucleophiles
Carboxylate Nucleophiles
Figure 6.Carboxylate Nucleophiles
Nitrogen Nucleophiles
A formidable challenge in asymmetric synthesis is the stereocontrolled construction of carbon-nitrogen bonds. Nitrogen nucleophiles such as alkylamines, azides, amides, imides, and N-heterocycles have all been employed in asymmetric allylic alkylation reactions.
Alkylamines Nucleophiles
Figure 7.Alkylamines Nucleophiles
Azide Nucleophiles
Figure 8.Azide Nucleophiles
Imide Nucleophiles
Figure 9.Imide Nucleophiles
Molybdenum-Catalyzed Reactions
The mechanism of the molybdenum-catalyzed AAA reaction is presumed to be distinctly different from the analogous Pd-catalyzed reaction, and in some cases, the levels of regio-, enantio-, and diastereoselectivity are enhanced relative to the palladium-catalyzed reaction. Trost and Dogra report the total synthesis of (–)-Δ9-trans-tetrahydrocannabinol, a psychomimetic of marijuana, utilizing a molybdenum catalyst.9 An overall yield of 30% of enantiomerically pure (–)-Δ9-trans-tetrahydrocannabinol (Figure 10).
Molybdenum-Catalyzed Reactions
Figure 10.Molybdenum-Catalyzed Reactions
References
1.
Trost BM, Fandrick D. 2007. The growing impact of asymmetric catalysis. Aldrichimica Acta. 40(59):
2.
Trost BM, Machacek MR, Aponick A. 2006. Predicting the Stereochemistry of Diphenylphosphino Benzoic Acid (DPPBA)-Based Palladium-Catalyzed Asymmetric Allylic Alkylation Reactions:  A Working Model. Acc. Chem. Res.. 39(10):747-760. https://doi.org/10.1021/ar040063c
3.
Trost BM. 2004. Asymmetric Allylic Alkylation, an Enabling Methodology. J. Org. Chem.. 69(18):5813-5837. https://doi.org/10.1021/jo0491004
4.
Trost BM, Crawley ML. 2003. Asymmetric Transition-Metal-Catalyzed Allylic Alkylations:  Applications in Total Synthesis. Chem. Rev.. 103(8):2921-2944. https://doi.org/10.1021/cr020027w
5.
Trost BM, Van Vranken DL. 1996. Asymmetric Transition Metal-Catalyzed Allylic Alkylations. Chem. Rev.. 96(1):395-422. https://doi.org/10.1021/cr9409804
6.
Trost BM. 1996. Designing a Receptor for Molecular Recognition in a Catalytic Synthetic Reaction:  Allylic Alkylation. Acc. Chem. Res.. 29(8):355-364. https://doi.org/10.1021/ar9501129
7.
Trost BM, Bunt RC. 1994. Asymmetric induction in allylic alkylations of 3-(acyloxy)cycloalkenes. J. Am. Chem. Soc.. 116(9):4089-4090. https://doi.org/10.1021/ja00088a059
8.
Helmchen G, Ernst M. A Novel Route to Iridoids: Enantioselective Syntheses of Isoiridomyrmecin and ?-Skytanthine. Synthesis. 2002(14 Special Issue): https://doi.org/10.1055/s-2002-34382
9.
Helmchen G, Ernst M. A Novel Route to Iridoids: Enantioselective Syntheses of Isoiridomyrmecin and ?-Skytanthine. Synthesis. 2002(41):4054-4056.
10.
Trost BM, Radinov R, Grenzer EM. 1997. Asymmetric Alkylation of ?-Ketoesters. J. Am. Chem. Soc.. 119(33):7879-7880. https://doi.org/10.1021/ja971523i
11.
Trost BM, Toste FD. 1999. Palladium-Catalyzed Kinetic and Dynamic Kinetic Asymmetric Transformation of 5-Acyloxy-2-(5H)-furanone. Enantioselective Synthesis of (?)-Aflatoxin B Lactone. J. Am. Chem. Soc.. 121(14):3543-3544. https://doi.org/10.1021/ja9844229
12.
Trost BM, Toste FD. 2003. Palladium Catalyzed Kinetic and Dynamic Kinetic Asymmetric Transformations of ?-Acyloxybutenolides. Enantioselective Total Synthesis of (+)-Aflatoxin B1and B2a. J. Am. Chem. Soc.. 125(10):3090-3100. https://doi.org/10.1021/ja020988s
13.
Trost BM, Crawley ML. 2002. 4-Aryloxybutenolides As ?Chiral Aldehyde? Equivalents:  An Efficient Enantioselective Synthesis of (+)-Brefeldin A. J. Am. Chem. Soc.. 124(32):9328-9329. https://doi.org/10.1021/ja026438b
14.
Trost BM, Crawley ML. 2004. A?Chiral Aldehyde? Equivalent as a Building Block Towards Biologically Active Targets. Chem. Eur. J.. 10(9):2237-2252. https://doi.org/10.1002/chem.200305634
15.
Trost BM, Kondo Y. 1991. An asymmetric synthesis of (+)-phyllanthocin. Tetrahedron Letters. 32(13):1613-1616. https://doi.org/10.1016/s0040-4039(00)74285-7
16.
Trost BM, Krische MJ, Radinov R, Zanoni G. 1996. On Asymmetric Induction in Allylic Alkylation via Enantiotopic Facial Discrimination. J. Am. Chem. Soc.. 118(26):6297-6298. https://doi.org/10.1021/ja960649x
17.
Trost BM, Pulley SR. 1995. Asymmetric Total Synthesis of (+)-Pancratistatin. J. Am. Chem. Soc.. 117(40):10143-10144. https://doi.org/10.1021/ja00145a038
18.
Trost BM, Joseph D, Hembre EJ. 2001. Asymmetric Induction of Conduritols via AAA Reactions: Synthesis of the Aminocyclohexitol of Hygromycin A. Chem.-Eur. J..(7):1619-1629.
19.
Trost BM, Van Vranken DL. 1993. A general synthetic strategy toward aminocyclopentitol glycosidase inhibitors. Application of palladium catalysis to the synthesis of allosamizoline and mannostatin A. J. Am. Chem. Soc.. 115(2):444-458. https://doi.org/10.1021/ja00055a013
20.
Trost BM, Van Vranken DL, Bingel C. 1992. A modular approach for ligand design for asymmetric allylic alkylations via enantioselective palladium-catalyzed ionizations. J. Am. Chem. Soc.. 114(24):9327-9343. https://doi.org/10.1021/ja00050a013
21.
Trost BM, Patterson DE. 1998. Enhanced Enantioselectivity in the Desymmetrization of Meso-Biscarbamates. J. Org. Chem.. 63(4):1339-1341. https://doi.org/10.1021/jo971746r
22.
Trost BM, Dogra K. 2007. Synthesis of (-)-?9-trans-Tetrahydrocannabinol:  Stereocontrol via Mo-Catalyzed Asymmetric Allylic Alkylation Reaction. Org. Lett.. 9(5):861-863. https://doi.org/10.1021/ol063022k