Merck
LC/MS Analysis of Melamine and Hydrolysis Products on Ascentis® Express HILIC
LC/MS Analysis of Melamine and Hydrolysis Products on Ascentis® Express HILIC application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express HILIC, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 5 cm × 2.1 mm
mobile phase component
Product No.
Description
Pricing
Ammonium formate
eluent additive for LC-MS, LiChropur, ≥99.0%
standard
Product No.
Description
Pricing
Melamine
99%
Ammelide
analytical standard
CONDITIONS
column
Ascentis Express HILIC, 5 cm x 2.1 mm I.D., 2.7 μm particles (53934-U)
mobile phase
5 mM ammonium formate in 95:5 (v/v) acetonitrile:water
flow rate
0.2 mL/min
column temp.
35 °C
detector
MS, ESI(+), full scan
injection
2 μL
sample
1 mg/L in mobile phase
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany