HPLC Analysis of Tryptic Digest on BIOshell A160 Peptide C18
![HPLC Analysis of Tryptic Digest on BIOshell A160 Peptide C18 application for HPLC](/deepweb/assets/sigmaaldrich/product/images/127/845/29ba17bb-3561-4525-9887-4a27b06e36fb/800/29ba17bb-3561-4525-9887-4a27b06e36fb.jpg)
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express Peptide ES-C18, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 15 cm × 2.1 mmstandard
Product No.
Description
Pricing
Casein from bovine milk
suitable for substrate for protein kinase (after dephosphorylation), purified powderCONDITIONS
column
BIOshell A160 Peptide C18, 15 cm x 2.1 mm I.D., 2.7 μm particles (66905-U)
mobile phase
[A] 0.1% formic acid in water; [B] 25:75, (0.4% formic acid in water):acetonitrile
gradient
0 to 70% B in 40 min.
flow rate
0.3 mL/min
column temp.
35 °C
detector
ESI(+), TOF
injection
2 μL
sample
10 pg/L in 0.1% formic acid
Description
Legal Information
null