HPLC Analysis of Synthetic Food Dyes on Ascentis® Express C18
Materials
analytical column
standard
CONDITIONS
column
Ascentis Express C18, 10 cm x 2.1 mm I.D., 2.7 μm particles (53823-U)
mobile phase
[A] 100 mM ammonium phosphate monobasic, pH 7.0 with ammonium hydroxide; [B] water; [C] acetonitrile
gradient
10% A constant; 85 to 25% B, 5 to 65% C in 6 min; 25% B, 65% C held for 5 min
flow rate
0.3 mL/min
column temp.
40 °C
detector
UV, 254 nm
injection
5 μL
sample
20 μg/mL each in water
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany