HPLC Analysis of Rabeprazole Enantiomers on Astec® CYCLOBOND I 2000 DMP
![HPLC Analysis of Rabeprazole Enantiomers on Astec® CYCLOBOND I 2000 DMP application for HPLC](/deepweb/assets/sigmaaldrich/product/images/100/867/481b480f-e210-41d1-b608-62070257fd86/800/481b480f-e210-41d1-b608-62070257fd86.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
CYCLOBOND I 2000 DMP, 25 cm x 4.6 mm I.D., 5 μm particles (20724AST)
mobile phase
[A] acetonitrile; [B] methanol; [C] acetic acid; [D] ammonium hydroxide; (95:5:0.4:0.1, A:B:C:D)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 230 nm
injection
5 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null