Merck
HPLC Analysis of Methyl Phenyl Sulfoxide Enantiomers on Astec® CHIROBIOTIC® TAG
HPLC Analysis of Methyl Phenyl Sulfoxide Enantiomers on Astec® CHIROBIOTIC® TAG application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® TAG Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
standard
Product No.
Description
Pricing
Methyl phenyl sulfoxide
≥97%
CONDITIONS
column
CHIROBIOTIC TAG, 25 cm x 4.6 mm I.D., 5 μm particles (14024AST)
mobile phase
methanol
flow rate
0.48 mL/min
column temp.
25 °C
detector
UV, 220 nm
injection
10 μL
sample
1 mg/mL in mobile phase
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC