Merck
HPLC Analysis of Insulin, Bovine on Discovery® C8
HPLC Analysis of Insulin, Bovine on Discovery® C8 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Discovery® C8 HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Discovery C8, 5 cm × 4.6 mm I.D., 5 μm particles (59353-U)
mobile phase
50 mM sodium phosphate, monobasic, 50 mM sodium sulfate, pH 3.0 (w/phosphoric acid), (A) +5% acetonitrile, (B) +50% acetonitrile
flow rate
1 mL/min
column temp.
35 °C
detector
UV, 220 nm
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany