HPLC Analysis of Diperodon Enantiomers on Astec® Cellulose DMP, LC/MS Conditions
![HPLC Analysis of Diperodon Enantiomers on Astec® Cellulose DMP, LC/MS Conditions application for HPLC](/deepweb/assets/sigmaaldrich/product/images/218/052/d8c248c6-c39a-4946-b8b7-ceaab1bfd032/800/d8c248c6-c39a-4946-b8b7-ceaab1bfd032.jpg)
CONDITIONS
column
Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 μm particles (51098AST)
mobile phase
[A] methanol; [B] ammonium formate; (100:0.1, A:B) (Note: 0.1 w/v ammonium formate, 0.1 g/100 mL)
flow rate
0.5 mL/min
column temp.
25 °C
detector
UV, 230 nm
injection
2 μL
sample
2 mg/mL in mobile phase