HPLC Analysis of Angiotensins on Discovery® BIO Wide Pore C18
Materials
analytical column
CONDITIONS
column
Discovery BIO Wide Pore C18, 15 cm × 4.6 mm I.D., 5 μm particles (568222-U)
mobile phase
[A] (10 mM NH4OAc, pH 7):(50% CH3CN in 20 mM NH4OAc, pH 7), 65:35 ; [B] (10 mM NH4OAc, pH 7):(50% CH3CN in 20 mM NH4OAc, pH 7), 25:75
gradient
0-100% B in 12.5 min
flow rate
1 mL/min
column temp.
ambient
detector
UV, 254 nm
injection
6 μL (10 μg each) in water
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany