GC Analysis of Organochlorine Pesticides in Water on SPB®-5 after SPME using 100 μm PDMS Fiber
Materials
SPME fiber
Product No.
Description
Pricing
SPME Fiber Assembly
100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, plain red hubCONDITIONS
sample/matrix
200 ppt each in 2 mL water
SPME fiber
polydimethylsiloxane, 100 μm (57300-U)
extraction
15 min immersed in water with rapid stirring
column
SPB-5, 15 m × 0.20 mm I.D., 0.20 μm (24165-U)
oven
120 °C (1 min) to 180 °C at 30 °C/min, then to 290 °C at 10 °C/min
inj. temp.
260 °C (splitless - closed 3 min)
detector
ECD, 300 °C
carrier gas
helium, 37 cm/sec (set at 120 °C)
Description
Legal Information
SPB is a registered trademark of Merck KGaA, Darmstadt, Germany