Merck
GC Analysis of Verbenone Enantiomers on Astec® CHIRALDEX G-TA
GC Analysis of Verbenone Enantiomers on Astec® CHIRALDEX™ G-TA suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIRALDEX G-TA Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.12 μm
standard
Product No.
Description
Pricing
(1S)-(−)-Verbenone
analytical standard
(1S)-(−)-Verbenone
94%
(1S)-(−)-Verbenone
≥93%
CONDITIONS
column
Astec CHIRALDEX G-TA, 30 m x 0.25 mm I.D., 0.12 μm (73033AST)
oven
120 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 psi
injection
1 μL, 80:1 split
sample
2.5 mg/mL
peak 1: R(+)-verbenone
peak 2: S(-)-verbenone
Description
Analysis Note
Can reverse elution order by using CHIRALDEX B-TA.
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC