Merck
GC Analysis of Stored Apples on 5% Phenyl Column after SPME using 100 μm PDMS Fiber
GC Analysis of Stored Apples on 5% Phenyl Column after SPME using 100 μm PDMS Fiber suitable for GC, application for SPME
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.50 μm
SPME fiber
Product No.
Description
Pricing
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB)
df 65 μm(PDMS/DVB, needle size 24 ga, for use with manual holder
related product
Product No.
Description
Pricing
SPME Fiber Holder
for use with manual sampling
SPME Fiber Holder
for use with CTC CombiPAL and Gerstel MPS2
standard
Product No.
Description
Pricing
Hexyl acetate
analytical standard
Butyl acetate
analytical standard
Isoamyl acetate
natural, ≥97%, FCC, FG
2-Methyl-1-butanol
≥99%
Hexyl 2-methylbutanoate
analytical standard
Hexyl hexanoate
analytical standard
CONDITIONS
sample/matrix
300-450g intact fruit in a 3 liter flask at ambient temp, exposed to an air flow of 25-30 mL/min
SPME fiber
polydimethylsiloxane, 100 μm film (57310-U)
desorption process
250°C for 90 sec
column
5% phenyl polysiloxane, 25m × 0.25 mm I.D., 0.34 μm film (Supelco equivalent, SLB-5ms, 28473-U)
oven
40 °C (1.5 min) to 250 °C at 50 °C/min, hold 1 min
inj. temp.
250 °C
detector
mass spectrometer, m/z = 40-300 (40 spectra/sec)
carrier gas
helium, 1.5 mL/min (44 cm/min)
sample
Apples
Description
Analysis Note
SPME can reduce sampling time and is complementary to rapid separation/detection techniques such as time compressed gas chromatography (TCGC) and time-of-flight mass spectrometry (TOF-MS), to permit analysis of dozens of compounds in a few minutes. Investigators have used SPME to monitor flavor volatiles in apple, strawberry, tomato, and many other fruits and vegetables. In this application, 65 μm PDMS/divinylbenzene (DVB) SPME fibers were used to capture volatiles emanating from Mutsu apples stored for 3-4 months at 0 °C.