Merck
GC Analysis of FAMEs in Human Plasma on SLB®-IL111
GC Analysis of FAMEs in Human Plasma on SLB®-IL111 suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-IL111 Capillary GC Column
L × I.D. 100 m × 0.25 mm, df 0.20 μm
related product
Product No.
Description
Pricing
Boron trifluoride-methanol solution
~10% (~1.3 M), derivatization grade (GC derivatization), LiChropur
standard
Product No.
Description
Pricing
F.A.M.E. Column Evaluation Mix
certified reference material, 1000 μg/mL each component in dichloromethane
CONDITIONS
column
SLB-IL111, 100 m x 0.25 mm I.D., 0.20 μm (29647-U)
oven
50 ºC (4 min), 8 ºC/min to 160 °C (25 min), 4 ºC/min to 220 ºC
inj. temp.
230 °C
detector
FID, 240 ºC
carrier gas
hydrogen, 40 cm/sec
injection
2 μL 10:1 split
sample
FAMEs from 500 μL human plasma transesterified in sodium methoxide/methanol + boron trifluoride
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany