Merck
  • A simple, rapid and sensitive RP-HPLC-UV method for the simultaneous determination of sorafenib & paclitaxel in plasma and pharmaceutical dosage forms: Application to pharmacokinetic study.
A simple, rapid and sensitive RP-HPLC-UV method for the simultaneous determination of sorafenib & paclitaxel in plasma and pharmaceutical dosage forms: Application to pharmacokinetic study.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2016-09-07)
Ismail Khan, Zafar Iqbal, Abad Khan, Muhammad Hassan, Fazle Nasir, Abida Raza, Lateef Ahmad, Amjad Khan, Muhammad Akhlaq Mughal
ABSTRACT
A simple, economical, fast, and sensitive RP-HPLC-UV method has been developed for the simultaneous quantification of Sorafenib and paclitaxel in biological samples and formulations using piroxicam as an internal standard. The experimental conditions were optimized and method was validated according to the standard guidelines. The separation of both the analytes and internal standard was achieved on Discovery HS C18 column (250mm×4.6mm, 5μm) using Acetonitrile and TFA (0.025%) in the ratio of (65:35V/V) as the mobile phase in isocratic mode at a flow rate of 1ml/min, with a wavelength of 245nm and at a column oven temperature of 25°Cin a short run time of 12min. The limits of detection (LLOD) were 5 and 10ng/ml while the limits of quantification (LLOQ) were 10 and 15ng/ml for sorafenib and paclitaxel, respectively. Sorafenib, paclitaxel and piroxicam (IS) were extracted from biological samples by applying acetonitrile as a precipitating and extraction solvent. The method is linear in the range of 15-20,000ng/ml for paclitaxel and 10-5000ng/ml for sorafenib, respectively. The method is sensitive and reliable by considering both of its intra-day and inter-day co-efficient of variance. The method was successfully applied for the quantification of the above mentioned drugs in plasma. The developed method will be applied towards sorafenib and paclitaxel pharmacokinetics studies in animal models.
Product Description
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Discovery® HS C18 Prep Guard Cartridge, 10 μm particle size, L × I.D. 1 cm × 21.2 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Discovery® HS C18 HPLC Column, 10 μm particle size, L × I.D. 25 cm × 21.2 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 21.2 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 2.1 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
Supelco
Discovery® HS C18 HPLC Column, 3 μm particle size, L × I.D. 7.5 cm × 2.1 mm
Supelco
Discovery® HS C18 HPLC Column, 3 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Discovery® HS C18 HPLC Column, 3 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Discovery® HS C18 HPLC Column, 3 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Discovery® HS C18 HPLC Column, 3 μm particle size, L × I.D. 5 cm × 2.1 mm
Supelco
Discovery® HS C18 HPLC Column, 3 μm particle size, L × I.D. 7.5 cm × 4.6 mm