Merck
  • Determination of perfluorinated alkylated substances in sediments and sediment core from the Gulf of Gdańsk, Baltic Sea.
Determination of perfluorinated alkylated substances in sediments and sediment core from the Gulf of Gdańsk, Baltic Sea.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering (2012-02-11)
Jerzy Falandysz, Paweł Rostkowski, Grażyna Jarzyńska, Jaromir J Falandysz, Sachi Taniyasu, Nobuyoshi Yamashita
ABSTRACT
Perfluorinated alkylated substances (PFAS) have been determined in surface sediments and sediment core from Gulf of Gdańsk, Baltic Sea. Perfluorooctanesulphonate (PFOS), perfluorohexanesulphonate (PFHxS), perfluorodecanoate (PFDA), perfluoronanoate (PFNA), perfluorooctanoate (PFOA), perfluoroheptanoate (PFHpA), perfluoroundecanote (PFUnDA), perfluorododecanoate (PFDoDA) and perfluorohexanoate (PFHxA) were quantified after isotopic dilution ((13)C(4) PFOS and (13)C(4) PFOA), liquid-liquid extractions by methanol and acetonitrile, cleanup by Envi-Carb, OasisWAX and Envi-Carb and final measurement by HPLC-MS/MS. PFOS, PFHxS, PFUnDA, PFDA, PFNA and PFOA were found in Baltic Sea sediments in concentrations exceeding the method limit of quantification (LOQ) of 2 pg/g. PFOS was detected in concentration up to 0.896 ng/g dry weight and PFHxS up to 0.326 ng/g dw, which shows on a weak pollution. PFOS (48-74%) or PFHxS (45-56%) dominated in PFAS composition of sediments surveyed. Supplemental materials are available for this article. Go to the publisher's online edition of the Journal of Environmental Science and Health, Part A to view the free supplemental file.
Product Description
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 1, pack of 100, suitable for EN 15662:2008 per BS
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 2, pack of 100, suitable for EN 15662:2008 per BS
Supelco
Supel QuE, PSA/ENVI-Carb Tube 2, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pk of 50, AOAC® 2007.01, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pack of 100, AOAC® 2007.01
Supelco
Supel QuE, PSA/ENVI-Carb Tube 1, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible