Merck
  • High-throughput analytical techniques for multiresidue, multiclass determination of 653 pesticides and chemical pollutants in tea--Part III: Evaluation of the cleanup efficiency of an SPE cartridge newly developed for multiresidues in tea.
High-throughput analytical techniques for multiresidue, multiclass determination of 653 pesticides and chemical pollutants in tea--Part III: Evaluation of the cleanup efficiency of an SPE cartridge newly developed for multiresidues in tea.
Journal of AOAC International (2013-09-05)
Guo-Fang Pang, Chun-Lin Fan, Qiao-Ying Chang, Yan Li, Jian Kang, Wen-Wen Wang, Jing Cao, Yan-Bing Zhao, Nan Li, Zeng-Yin Li, Zong-Mao Chen, Feng-Jian Luo, Zheng-Yun Lou
ABSTRACT
A comparative study was conducted over three stages on the cleanup efficiency of SPE cartridge Cleanert TPT, newly developed for multigroups of pesticide residues in tea. In Stage I, different SPE cartridges C18, graphite carbon black (GCB), primary secondary amine (PSA), and amino (NH2) were purchased and combined into 12 different sequences. Through the comparative test on cleanup efficiency of 84 representative pesticides in tea, Envi-Carb GCB + PSA with a good cleanup effect was selected. In Stage II, GC/MS test results from the comparative study of the extraction efficiency of 201 pesticides spiked into green tea and Woolong tea with Cleanert TPT and Envi-Carb + PSA SPE showed that average recoveries fell within 70-110% and RSD <20% for 193 and 184 pesticides, respectively, for green tea, accounting for 96.0 and 91.0% of the total number, respectively. GC/MS/MS test results also found 193 and 184 pesticides, respectively, meeting the recovery and RSD conditions, accounting for 96.0 and 91.5%, respectively, of the total number. For Woolong tea samples, GC/MS results showed that with Cleanert TPT and Envi-Carb + PSA SPE for cleanup, there were 192 and 177 pesticides, respectively, meeting the conditions, accounting for 95.5 and 88.1% of the total number, respectively. GC/MS/MS results demonstrated that there were 195 and 184 pesticides, respectively, meeting the conditions, accounting for 97.0 and 91.5% of the total number, respectively. It was seen that Cleanert TPT was superior to Envi-Carb + PSA in cleanup efficiency, whether for green or Woolong tea samples, or GC/MS or GC/MS/MS determination. In Stage III, 61104 results of the average content value of pesticides and RSD (two teas xtwo Youden pair concentrations x two kinds of SPE cartridges x two instruments x 19 tests x 201 pesticides) were derived from the 19 times stability tests over 3 months by paralleling three samples every 5 days via two instruments with two kinds of SPE cartridges for cleanup, respectively, against Youden Pair samples of the 201 incurred pesticides from green and Woolong teas. The statistical analysis found that detected values from the target pesticides of the incurred Youden pair samples showed no marked differences with cleanup by either Cleanert TPT or Envi-Carb + PSA, whether for green or Woolong tea, or G/IMS or G/IM/IMS. The test results using the two aforementioned kinds of SPE cleanup for above 93% pesticides had a tolerance less than 15%, which testifies that both cartridge cleanups met the requirement for pesticide residue analysis.
Product Description
Supelco
Supel QuE, PSA/ENVI-Carb Tube 2, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pk of 50, AOAC® 2007.01, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb Tube 1, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 1, pack of 100, suitable for EN 15662:2008 per BS
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pack of 100, AOAC® 2007.01
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 2, pack of 100, suitable for EN 15662:2008 per BS