Merck
  • Progesterone and beta-estradiol influence the cell density-dependent distribution of cyclic nucleotides across the cell membrane of human C4-I cells (squamous carcinoma of the uterine cervix) by a non-genomic mechanism.
Progesterone and beta-estradiol influence the cell density-dependent distribution of cyclic nucleotides across the cell membrane of human C4-I cells (squamous carcinoma of the uterine cervix) by a non-genomic mechanism.
Anticancer research (1995-09-01)
A Orbo, R Jaeger, K E Kjørstad, G Sager
ABSTRACT
The ratio between extracellular levels of cGMP and cAMP (cGMPex/cAMPex) has been proposed as diagnostic tool in many forms of malignancies. In vitro and in vivo studies have shown that sex steroids effect extracellular levels of cyclic nucleotides. Cyclic changes of these hormones in premenopausal women may disturb the interpretation of the diagnostic marker. C4-I cells grew in the presence of beta-estradiol and progesterone in a chemically defined medium. Cells were sampled during the logarithmic growth phase. Cyclic nucleotide levels were determined by RIA. Receptor status was evaluated by immunocytochemistry. Progesterone increased the cGMPex/cAMPex at all cell densities tested. This effect resulted from increased cGMP and reduced cAMP extrusion. Estradiol had no clear effect on cGMPex/cAMPex even when inhibition of cAMP extrusion was observed at low cell density. Receptors for steroids were not detectable. Sex steroids interact with cyclic nucleotides in C4-I cells in a non-genomic manner.
Product Description
Supelco
ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Supelco
ORBO 65P (OVS) Amberlite XAD®-4 specially cleaned, 160/80 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS), pkg of 10 ea
Supelco
ORBO 52 Small Activated Silica Gel (20/40) 150/75 mg, pk 50, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 24 2-HMP on Amberlite XAD®-2 (20/40), 150/75 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 25 ea
Supelco
ORBO 92 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group Vinyl acetate
Supelco
ORBO 43 Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 80 Isocyanate Filter, with cassettes: no, filter O.D. 37 mm, pkg of 25 ea
Supelco
ORBO 402 Tenax® TA specially treated (35/60), 100/50 mg, W,W,W separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 609 Amberlite XAD®-2 (20/50) 400/200 mg, W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
Supelco
ORBO 53 Activated Silica Gel (20/40) specially cleaned, 400/200 mg with Glass Fiber Filters, GFF,F,F separators, O.D. × L 7 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 502 Activated Silica Gel (20/45) 100/50 mg, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 49P (OVS) Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 270/140 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS) Tube, pkg of 10 ea
Supelco
ORBO 554 H2SO4 on Silica Gel (20/40) 150/75 mg, W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 80 Coated Filter, with cassettes unassembled
Supelco
ORBO 23 2-HMP on Amberlite XAD®-2 (20/40), 120/60 mg, W,W,W separators, O.D. × L 6 mm × 85 mm, pkg of 25 ea