Merck
  • Rapid non-genomic and concentration-dependent effects of progesterone in c4-I cells on the proposed tumor-marker - ratio between extracellular cgmp and cAMP levels.
Rapid non-genomic and concentration-dependent effects of progesterone in c4-I cells on the proposed tumor-marker - ratio between extracellular cgmp and cAMP levels.
International journal of oncology (1995-06-01)
A Orbo, K Kjorstad, R Jaeger, G Sager
ABSTRACT
The extracellular cGMP levels or the ratio between extracellular levels of cGMP and cAMP (cGMP(ex)/cAMP(ex)) have been proposed as tumor marker for premalignant and malignant diseases of the uterine cervix. More than 50% of cervical cancers occur in premenopausal women and detailed information about hormonal and drug effects on the extracellular levels of cyclic nucleotides is of importance. In the present study we have investigated the effect of progesterone (0.1-100 mu M), theophylline (1-1000 mu M), probenecid (0.1-100 mu M) and verapamil (0.1-100 mu M) on cGMP(ex)/cAMP(ex) of C4-I cells (a human cell line derived from a squamous carcinoma of the uterine cervix). Within 30 min progesterone caused a concentration-dependent elevation of cGMP(ex)/cAMP(ex), whereas the other compounds had no marked effect. Identical results were obtained for C4-I cells in monolayer and in suspension. The effects were explained by the observation that progesterone stimulated cGMP efflux, but inhibited the cAMP efflux. The other compounds inhibited the export of both nucleotides to a similar degree. The present data suggest that progesterone affects the export of cyclic nucleotides in non-genomic manner and may hamper the interpretation of cGMP(ex)/cAMP(ex) in the luteal phase in premenopausal women with cancer of the uterine cervix.
Product Description
Supelco
ORBO 609 Amberlite XAD®-2 (20/50) 400/200 mg, W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
Supelco
ORBO 402 Tenax® TA specially treated (35/60), 100/50 mg, W,W,W separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 23 2-HMP on Amberlite XAD®-2 (20/40), 120/60 mg, W,W,W separators, O.D. × L 6 mm × 85 mm, pkg of 25 ea
Supelco
ORBO 49P (OVS) Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 270/140 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS) Tube, pkg of 10 ea
Supelco
ORBO 502 Activated Silica Gel (20/45) 100/50 mg, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 53 Activated Silica Gel (20/40) specially cleaned, 400/200 mg with Glass Fiber Filters, GFF,F,F separators, O.D. × L 7 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 554 H2SO4 on Silica Gel (20/40) 150/75 mg, W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 80 Coated Filter, with cassettes unassembled
Supelco
ORBO 65P (OVS) Amberlite XAD®-4 specially cleaned, 160/80 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS), pkg of 10 ea
Supelco
ORBO 92 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group Vinyl acetate
Supelco
ORBO 43 Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 24 2-HMP on Amberlite XAD®-2 (20/40), 150/75 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 25 ea
Supelco
ORBO 52 Small Activated Silica Gel (20/40) 150/75 mg, pk 50, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 80 Isocyanate Filter, with cassettes: no, filter O.D. 37 mm, pkg of 25 ea
Supelco
ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)