Merck
Rapid Multi-Residue Analysis of Pesticides in Pulses by LC-MS/MS.
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan (2016-10-28)
Keiko Iwakoshi, Kenji Otsuka, Yasuhiro Tamura, Sanae Tomizawa, Tamako Masubuchi, Yumiko Yamaki, Yukiko Nakagawa, Ryoko Masuda, Shota Suto, Yoshie Kokaji, Tetsuya Shindo, Ichiro Takano
ABSTRACT
Rapid multi-residue analysis of pesticides in pulses was developed using LC-MS/MS. Pesticide residues in 5 g of homogenized pulses were extracted with 30 mL of acetonitrile and salted out with 4 g of anhydrous magnesium sulfate and 2 g of sodium chloride in the presence of citrate buffer in a disposable tube. The resulting residues were extracted with 30 mL of acetonitrile, and co-extractives were removed on a handmade four-layer column, consisting of a layer of Z-Sep/C18 (20 mg/50 mg) dry particles on top of a three-layer, custom-made (pre-packed) column (lower bed: 60 mg of PSA, middle bed: 30 mg of GC, and top bed: 60 mg of C18) packed in a 10 mm internal diameter polypropylene column (3 mL). The developed method showed good recoveries of pesticides in soybean, lentil, white kidney bean and garbanzo. According to the method validation guideline of the Ministry of Health, Labour and Welfare of Japan, recovery tests were conducted in soybeans fortified with 107 kinds of pesticides at the levels of 0.01 and 0.1 μg/g, respectively. At each concentration 2 samples were extracted on 5 separate days. Pesticides in the test solution were determined by LC-MS/MS using scheduled MRM. As regards the trueness of this method for 107 pesticides in soybeans, 97 pesticides were in the range of 70-120% with satisfactory repeatability and within-run reproducibility. This new method is expected to be applicable for routine examination of pesticide residues in soybeans.
Product Description
Supelco
Supel QuE, Z-Sep+ Tube, centrifuge tube volume 2 mL, pack of 100 ea
Supelco
Supel QuE, Z-Sep Bulk
Supelco
Supel QuE, Z-Sep/C18 Tube, centrifuge tube volume 2 mL, pack of 100
Supelco
Supel QuE, Z-Sep Tube, centrifuge tube volume 2 mL, pack of 100 ea
Supelco
Supel QuE, Z-Sep+ Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, Z-Sep Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, Z-Sep/C18 Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, Z-Sep+ with MgSO4 Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, Z-Sep+ Bulk, pkg of 20 g