Merck
  • Circadian coordination of ATP release in the urothelium via connexin43 hemichannels.
Circadian coordination of ATP release in the urothelium via connexin43 hemichannels.
Scientific reports (2018-02-02)
Atsushi Sengiku, Masakatsu Ueda, Jin Kono, Takeshi Sano, Nobuyuki Nishikawa, Sumihiro Kunisue, Kojiro Tsujihana, Louis S Liou, Akihiro Kanematsu, Shigeki Shimba, Masao Doi, Hitoshi Okamura, Osamu Ogawa, Hiromitsu Negoro
ABSTRACT
Day-night changes in the storage capacity of the urinary bladder are indispensable for sound sleep. Connexin 43 (Cx43), a major gap junction protein, forms hemichannels as a pathway of ATP in other cell types, and the urinary bladder utilizes ATP as a mechanotransduction signals to modulate its capacity. Here, we demonstrate that the circadian clock of the urothelium regulates diurnal ATP release through Cx43 hemichannels. Cx43 was expressed in human and mouse urothelium, and clock genes oscillated in the mouse urothelium accompanied by daily cycles in the expression of Cx43 and extracellular ATP release into the bladder lumen. Equivalent chronological changes in Cx43 and ATP were observed in immortalized human urothelial cells, but these diurnal changes were lost in both arrhythmic Bmal1-knockout mice and in BMAL1-knockdown urothelial cells. ATP release was increased by Cx43 overexpression and was decreased in Cx43 knockdown or in the presence of a selective Cx43 hemichannel blocker, which indicated that Cx43 hemichannels are considered part of the components regulating ATP release in the urothelium. Thus, a functional circadian rhythm exists in the urothelium, and coordinates Cx43 expression and function as hemichannels that provide a direct pathway of ATP release for mechanotransduction and signalling in the urothelium.
Sigma-Aldrich
Carbenoxolone disodium salt, ≥98%