Merck
G-quadruplex-binding small molecules ameliorate
EMBO molecular medicine (2017-11-09)
Roberto Simone, Rubika Balendra, Thomas G Moens, Elisavet Preza, Katherine M Wilson, Amanda Heslegrave, Nathan S Woodling, Teresa Niccoli, Javier Gilbert-Jaramillo, Samir Abdelkarim, Emma L Clayton, Mica Clarke, Marie-Therese Konrad, Andrew J Nicoll, Jamie S Mitchell, Andrea Calvo, Adriano Chio, Henry Houlden, James M Polke, Mohamed A Ismail, Chad E Stephens, Tam Vo, Abdelbasset A Farahat, W David Wilson, David W Boykin, Henrik Zetterberg, Linda Partridge, Selina Wray, Gary Parkinson, Stephen Neidle, Rickie Patani, Pietro Fratta, Adrian M Isaacs
ABSTRACT
Intronic GGGGCC repeat expansions in
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Purmorphamine, ≥98% (HPLC)