- Molecular architecture of the N-type ATPase rotor ring from
Molecular architecture of the N-type ATPase rotor ring from
EMBO reports (2017-03-12)
Sarah Schulz, Martin Wilkes, Deryck J Mills, Werner Kühlbrandt, Thomas Meier
PMID28283532
ABSTRACT
The genome of the highly infectious bacterium
Sigma-Aldrich
Monoclonal Anti-polyHistidine antibody produced in mouse, clone HIS-1, ascites fluid