Merck
  • Smad4 restricts differentiation to promote expansion of satellite cell derived progenitors during skeletal muscle regeneration.
Smad4 restricts differentiation to promote expansion of satellite cell derived progenitors during skeletal muscle regeneration.
eLife (2016-11-20)
Nicole D Paris, Andrew Soroka, Alanna Klose, Wenxuan Liu, Joe V Chakkalakal
ABSTRACT
Skeletal muscle regenerative potential declines with age, in part due to deficiencies in resident stem cells (satellite cells, SCs) and derived myogenic progenitors (MPs); however, the factors responsible for this decline remain obscure. TGFβ superfamily signaling is an inhibitor of myogenic differentiation, with elevated activity in aged skeletal muscle. Surprisingly, we find reduced expression of
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution