- Deletional tolerance prevents AQP4-directed autoimmunity in mice.
Deletional tolerance prevents AQP4-directed autoimmunity in mice.
European journal of immunology (2017-01-07)
Anna-Lena Vogel, Benjamin Knier, Katja Lammens, Sudhakar Reddy Kalluri, Tanja Kuhlmann, Jeffrey L Bennett, Thomas Korn
PMID28058717
ABSTRACT
Neuromyelitis optica (NMO) is an autoimmune disorder of the central nervous system (CNS) mediated by antibodies to the water channel protein AQP4 expressed in astrocytes. The contribution of AQP4-specific T cells to the class switch recombination of pathogenic AQP4-specific antibodies and the inflammation of the blood-brain barrier is incompletely understood, as immunogenic naturally processed T-cell epitopes of AQP4 are unknown. By immunizing Aqp4