Merck
SAP97 Binding Partner CRIPT Promotes Dendrite Growth
eNeuro (2017-12-09)
Lei Zhang, Angela Marie Jablonski, Jelena Mojsilovic-Petrovic, Hua Ding, Steven Seeholzer, Ian Paterson Newton, Inke Nathke, Rachael Neve, JinBin Zhai, Yuan Shang, Mingjie Zhang, Robert Gordon Kalb
ABSTRACT
The dendritic tree is a key determinant of neuronal information processing. In the motor system, the dendritic tree of spinal cord neurons undergoes dramatic remodeling in an activity-dependent manner during early postnatal life. This leads to the proper segmental spinal cord connectivity that subserves normal locomotor behavior. One molecular system driving the establishment of dendrite architecture of mammalian motor neurons relies on AMPA receptors (AMPA-Rs) assembled with the GluA1 subunit, and this occurs in an NMDA receptor (NMDA-R)-independent manner. The dendrite growth promoting activity of GluA1-containing AMPA-Rs depends on its intracellular binding partner, SAP97, and SAP97's PDZ3 domain. We show here that cysteine-rich interactor of PDZ3 (CRIPT) is a
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution