Merck
  • The Significance of the Sulfatase Pathway for Local Estrogen Formation in Endometrial Cancer.
The Significance of the Sulfatase Pathway for Local Estrogen Formation in Endometrial Cancer.
Frontiers in pharmacology (2017-07-12)
Maša Sinreih, Tamara Knific, Maja Anko, Neli Hevir, Katja Vouk, Aleš Jerin, Snježana Frković Grazio, Tea Lanišnik Rižner
ABSTRACT
Endometrial cancer (EC) is the most common estrogen-dependent gynecological malignancy in the developed World. To investigate the local formation of estradiol (E2), we first measured the concentrations of the steroid precursor androstenedione (A-dione) and the most potent estrogen, E2, and we evaluated the metabolism of A-dione, estrone-sulfate (E1-S), and estrone (E1) in cancerous and adjacent control endometrium. Furthermore, we studied expression of the key genes for estradiol formation via the aromatase and sulfatase pathways. A-dione and E2 were detected in cancerous and adjacent control endometrium. In cancerous endometrium, A-dione was metabolized to testosterone, and no E2 was formed. Both, E1-S and E1 were metabolized to E2, with increased levels of E2 seen in cancerous tissue. There was no significant difference in expression of the key genes of the aromatase (