- Gastrodin Inhibits Store-Operated Ca
Gastrodin Inhibits Store-Operated Ca
Frontiers in pharmacology (2017-05-11)
Changbo Zheng, Chun-Yin Lo, Zhaoyue Meng, Zhichao Li, Mingkui Zhong, Peng Zhang, Jun Lu, Zhaoxiang Yang, Fuman Yan, Yunting Zhang, Yu Huang, Xiaoqiang Yao
PMID28487655
ABSTRACT
Cardiac hypertrophy is a major risk factor for heart failure, which are among the leading causes of human death. Gastrodin is a small molecule that has been used clinically to treat neurological and vascular diseases for many years without safety issues. In the present study, we examined protective effect of gastrodin against cardiac hypertrophy and explored the underlying mechanism. Phenylephrine and angiotensin II were used to induce cardiac hypertrophy in a mouse model and a cultured cardiomyocyte model. Gastrodin was found to alleviate the cardiac hypertrophy in both models. Mechanistically, gastrodin attenuated the store-operated Ca